1-Methyl-3-indolemethylamine, 96%
1-Methyl-3-indolemethylamine, 96%
1-Methyl-3-indolemethylamine, 96%
Thermo Scientific Chemicals

1-Methyl-3-indolemethylamine, 96%

CAS: 19293-60-8 | C10H12N2 | 160.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H34418.06
also known as H34418-06
Price (USD)
635.65
Online Exclusive
706.00
Save 70.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
635.65
Online Exclusive
706.00
Save 70.35 (10%)
Each
Chemical Identifiers
CAS19293-60-8
IUPAC Name1-(1-methyl-1H-indol-3-yl)methanamine
Molecular FormulaC10H12N2
InChI KeyNOFZMDGMQKRLIV-UHFFFAOYSA-N
SMILESCN1C=C(CN)C2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive Index1.6130-1.6190 @ 20?C
FormLiquid
Appearance (Color)Clear colorless to pale yellow
Assay (Non-aqueous acid-base Titration)≥95.0 to ≤105.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only