1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride, 95%
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride, 95%
1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride, 95%
Thermo Scientific Chemicals

1-(5-Trifluoromethyl-2-pyridyl)-1H-pyrazole-4-sulfonyl chloride, 95%

CAS: 1006441-36-6 | C9H5ClF3N3O2S | 311.663 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34430.MD
also known as H34430-MD
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.50
Save 11.85 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
69.65
Special offer
Online exclusive
Ends: 30-Jun-2026
81.50
Save 11.85 (15%)
Each
Chemical Identifiers
CAS1006441-36-6
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)82.5-91.5?C
Appearance (Color)White
Assay (GC)≥94.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only