6-Methoxypyridine-2-carboxaldehyde, 96%
6-Methoxypyridine-2-carboxaldehyde, 96%
6-Methoxypyridine-2-carboxaldehyde, 96%
6-Methoxypyridine-2-carboxaldehyde, 96%
Thermo Scientific Chemicals

6-Methoxypyridine-2-carboxaldehyde, 96%

CAS: 54221-96-4 | C7H7NO2 | 137.138 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34432.06
also known as H34432-06
Price (USD)
215.65
Special offer
Online exclusive
Ends: 30-Jun-2026
253.00
Save 37.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
215.65
Special offer
Online exclusive
Ends: 30-Jun-2026
253.00
Save 37.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS54221-96-4
IUPAC Name6-methoxypyridine-2-carbaldehyde
Molecular FormulaC7H7NO2
InChI KeyYDNWTNODZDSPNZ-UHFFFAOYSA-N
SMILESCOC1=CC=CC(C=O)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear yellow to orange to brown
Refractive Index1.5280-1.5340 @ 20?C
Free acid (titration)≤2%
Assay (GC)≥95.0%
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only