4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%
4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%
Thermo Scientific Chemicals

4-(2-Oxo-1-pyrrolidinyl)benzenesulfonyl chloride, 97%

CAS: 112539-09-0 | C10H10ClNO3S | 259.704 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
5 g
Catalog number H34441.MD
also known as H34441-MD
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.40
Save 8.75 (14%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
51.65
Special offer
Online exclusive
Ends: 30-Jun-2026
60.40
Save 8.75 (14%)
Each
Chemical Identifiers
CAS112539-09-0
IUPAC Name4-(2-oxopyrrolidin-1-yl)benzene-1-sulfonyl chloride
Molecular FormulaC10H10ClNO3S
InChI KeyOUAVTZYGHGCCCB-UHFFFAOYSA-N
SMILESClS(=O)(=O)C1=CC=C(C=C1)N1CCCC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or needles
Appearance (Color)White
Assay (Titration ex Chloride)≥96.0 to ≤104.0%
Melting Point (clear melt)104.0-113.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only