4-Trifluoromethylbenzoic anhydride, 97%
4-Trifluoromethylbenzoic anhydride, 97%
4-Trifluoromethylbenzoic anhydride, 97%
Thermo Scientific Chemicals

4-Trifluoromethylbenzoic anhydride, 97%

CAS: 25753-16-6 | C16H8F6O3 | 362.227 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H34480.03
also known as H34480-03
Price (USD)
34.10
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
34.10
Each
Chemical Identifiers
CAS25753-16-6
IUPAC Name4-(trifluoromethyl)benzoyl 4-(trifluoromethyl)benzoate
Molecular FormulaC16H8F6O3
InChI KeyFNAWJOBKLWLHTA-UHFFFAOYSA-N
SMILESFC(F)(F)C1=CC=C(C=C1)C(=O)OC(=O)C1=CC=C(C=C1)C(F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
FormCrystals or powder or crystalline powder
Assay (GC)≥96.0%
Melting Point (clear melt)127.0-133.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only