1-Eicosanethiol, 98%
1-Eicosanethiol, 98%
1-Eicosanethiol, 98%
Thermo Scientific Chemicals

1-Eicosanethiol, 98%

CAS: 13373-97-2 | C20H42S | 314.616 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H34498.MD
also known as H34498-MD
Price (USD)
76.65
Online Exclusive
85.30
Save 8.65 (10%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
76.65
Online Exclusive
85.30
Save 8.65 (10%)
Each
Chemical Identifiers
CAS13373-97-2
IUPAC Nameicosane-1-thiol
Molecular FormulaC20H42S
InChI KeyYYHYWOPDNMFEAV-UHFFFAOYSA-N
SMILESCCCCCCCCCCCCCCCCCCCCS
View more
SpecificationsSpecification SheetSpecification Sheet
FormFused solid, crystals or crystalline powder
Melting Point (clear melt)37-43?C
Assay (GC)≥97.5%
Appearance (Color)White
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Eicosanethiol is used as an organic chemical synthesis intermediate.

Notes
Air and moisture sensitive. Incompatible with heat.
RUO – Research Use Only

General References:

  1. Y.Contreras; A.J.Muscat. Passivation of InSb(100) with 1-eicosanethiol self-assembled monolayers. Solid State Phenomena. 2014, 219,59-62.
  2. Masashi Watanabe; Ryou Hatano; Chiho Koizumi. Spontaneous formation of interdigitated array pattern in wrinkled gold films deposited on poly(dimethylsiloxane) elastomer. Polymer Journal. 2015, 47,320-327.