2,4-Dimethyl-1,3-pentadiene, 95%
2,4-Dimethyl-1,3-pentadiene, 95%
2,4-Dimethyl-1,3-pentadiene, 95%
Thermo Scientific Chemicals

2,4-Dimethyl-1,3-pentadiene, 95%

CAS: 1000-86-8 | C7H12 | 96.173 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H35300.06
also known as H35300-06
Price (USD)
82.65
Online Exclusive
91.90
Save 9.25 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
82.65
Online Exclusive
91.90
Save 9.25 (10%)
Each
Chemical Identifiers
CAS1000-86-8
IUPAC Name2,4-dimethylpenta-1,3-diene
Molecular FormulaC7H12
InChI KeyCMSUNVGIWAFNBG-UHFFFAOYSA-N
SMILESCC(C)=CC(C)=C
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥94.0%
Appearance (Color)Clear colorless
FormLiquid
Refractive Index1.4385-1.4425 @ 20°C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only