2-Chloro-6-nitrobenzothiazole, 95%
2-Chloro-6-nitrobenzothiazole, 95%
Thermo Scientific Chemicals

2-Chloro-6-nitrobenzothiazole, 95%

CAS: 2407-11-6 | C7H3ClN2O2S | 214.62 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Catalog number H35525.09
also known as H35525-09
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)
198.65
Special offer
Online exclusive
Ends: 30-Jun-2026
233.00
Save 34.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS2407-11-6
IUPAC Name2-chloro-6-nitro-1,3-benzothiazole
Molecular FormulaC7H3ClN2O2S
InChI KeyKUCSJGBXJBQHNI-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C2N=C(Cl)SC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥94.0%
Melting Point (clear melt)185.0-197.0?C
Appearance (Color)Pale cream to pale yellow
FormPowder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only