2-Chloro-5-nitrobenzophenone, 98%
2-Chloro-5-nitrobenzophenone, 98%
2-Chloro-5-nitrobenzophenone, 98%
Thermo Scientific Chemicals

2-Chloro-5-nitrobenzophenone, 98%

CAS: 34052-37-4 | C13H8ClNO3 | 261.661 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number H35800.22
also known as H35800-22
Price (USD)
157.65
Special offer
Online exclusive
Ends: 28-Dec-2026
185.00
Save 27.35
Each
Quantity:
100 g
Request bulk or custom format
Price (USD)
157.65
Special offer
Online exclusive
Ends: 28-Dec-2026
185.00
Save 27.35
Each
Chemical Identifiers
CAS34052-37-4
IUPAC Name(2-chloro-5-nitrophenyl)(phenyl)methanone
Molecular FormulaC13H8ClNO3
InChI KeyHRPHZUAPQWJPCZ-UHFFFAOYSA-N
SMILES[O-][N+](=O)C1=CC=C(Cl)C(=C1)C(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥97.5%
Appearance (Color)Pale yellow to orange or pale cream to cream to brown
FormPowder
Melting Point (clear melt)80.5-86.5?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only