2-Iodoresorcinol, 97%
2-Iodoresorcinol, 97%
2-Iodoresorcinol, 97%
Thermo Scientific Chemicals

2-Iodoresorcinol, 97%

CAS: 41046-67-7 | C6H5IO2 | 236.008 g/mol
Have Questions?
Catalog NumberQuantity
H35941.03
also known as H35941-03
1 g
Catalog number H35941.03
also known as H35941-03
Price (USD)
130.00
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
130.00
Each
Chemical Identifiers
CAS41046-67-7
IUPAC Name2-iodobenzene-1,3-diol
Molecular FormulaC6H5IO2
InChI KeyBNJXHRMYHDWZKL-UHFFFAOYSA-N
SMILESOC1=CC=CC(O)=C1I
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale pink or pale yellow to orange or pale cream to pale brown
FormPowder
Assay (HPLC)≥96.0%
Melting Point (clear melt)100-111?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only