1,3,5-Benzenetriboronic acid tris(pinacol) ester, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Benzenetriboronic acid tris(pinacol) ester, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36045.03
also known as H36045-03
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.10
Save 12.45
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
70.65
Special offer
Online exclusive
Ends: 28-Dec-2026
83.10
Save 12.45
Each
Chemical Identifiers
CAS365564-05-2
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)≥96.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to cream or pale brown
1. Fabricated a 2D COF which when deposited onto a Gold electrode increases the hydrogen evolution reaction(HER) activity by up to three times compared with bare Gold.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.