2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%
2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%
Thermo Scientific Chemicals

2,4,6-Tris(3-pyridyl)-1,3,5-triazine, 97%

CAS: 42333-76-6 | C18H12N6 | 312.34 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36052.06
also known as H36052-06
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
278.65
Online Exclusive
309.00
Save 30.35 (10%)
Each
Chemical Identifiers
CAS42333-76-6
SpecificationsSpecification SheetSpecification Sheet
FormPowder
Assay (Non-aqueous acid-base Titration)≥96.0 to ≤104.0%
Melting Point (DSC)306.0-316.0?C
Appearance (Color)Brown to gray
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only