Triphenylcarbenium tetrakis(pentafluorophenyl)borate, 97%
Thermo Scientific Chemicals

Triphenylcarbenium tetrakis(pentafluorophenyl)borate, 97%

Have Questions?
Catalog NumberQuantity
H36154.03
also known as H36154-03
1 g
Catalog number H36154.03
also known as H36154-03
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
171.65
Online exclusive
191.00
Save 19.35
Each
Chemical Identifiers
CAS136040-19-2
IUPAC Nametetrakis(2,3,4,5,6-pentafluorophenyl)boranuide; triphenylmethylium
Molecular FormulaC43H15BF20
InChI KeyTZOSNOQHGGONMD-UHFFFAOYSA-N
SMILESC1=CC=C(C=C1)[C+](C1=CC=CC=C1)C1=CC=CC=C1.FC1=C(F)C(F)=C(C(F)=C1F)[B-](C1=C(F)C(F)=C(F)C(F)=C1F)(C1=C(F)C(F)=C(F)C(F)=C1F)C1=C(F)C(F)=C(F)C(F)=C1F
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormPowder
Melting Point (clear melt)174.0-183.0?C
Assay (Aqueous acid-base Titration)≥96.0 to ≤104.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.