4-tert-Butyl-2,6-diformylphenol, 96%
Thermo Scientific Chemicals

4-tert-Butyl-2,6-diformylphenol, 96%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36238.03
also known as H36238-03
Price (USD)
48.65
Online Exclusive
54.30
Save 5.65 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
48.65
Online Exclusive
54.30
Save 5.65 (10%)
Each
Chemical Identifiers
CAS84501-28-0
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)white or yellow to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
1. Used as a key intermediate for the production of a BODIPY chemodosimetric sensor for cyanide ions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.