Lithium hydroxide monohydrate, battery grade
Lithium hydroxide monohydrate, battery grade
Lithium hydroxide monohydrate, battery grade
Lithium hydroxide monohydrate, battery grade
Thermo Scientific Chemicals

Lithium hydroxide monohydrate, battery grade

CAS: 1310-66-3 | H3LiO2 | 41.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number H36379.0B
also known as H36379-0B
Price (USD)
411.65
Online Exclusive
457.00
Save 45.35 (10%)
Each
Quantity:
1000 g
Request bulk or custom format
Price (USD)
411.65
Online Exclusive
457.00
Save 45.35 (10%)
Each
Chemical Identifiers
CAS1310-66-3
IUPAC Namelithium(1+) hydrate hydroxide
Molecular FormulaH3LiO2
InChI KeyGLXDVVHUTZTUQK-UHFFFAOYSA-M
SMILES[Li+].O.[OH-]
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
FormCrystalline powder
Assay (special)≥54.0% (as LiOH)
Impurity contentSodium + Potassium, Na + K: ≤3000ppm (≤0.3%)
Chloride content≤500ppm (≤0.05%)
View more
Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Lithium hydroxide monohydrate is used in preparation of other lithium salts, as a catalyst in the production of alkyd resins, in esterifications. It is also used in the production of lithium soaps, greases, sulfonates and electric storage batteries.

Solubility
Soluble in water (216 mg/ml at 20°C), ethanol (freely), and alcohol (slightly).

Notes
Air sensitive and hygroscopic. Incompatible with oxidizing agents and acids.
RUO – Research Use Only

General References:

  1. Yangsoo Kim; Kyu Yong Choi; Thomas A.Chamberlin. Kinetics of melt transesterification of diphenyl carbonate and bisphenol A to polycarbonate with lithium hydroxide monohydrate catalyst. Ind. Eng. Chem. Res. 1992, 31, (9),2118-2127
  2. Kersti Hermansson. From cluster to crystal: Ab initio calculations of the OH- frequency in lithium hydroxide monohydrate. Chemical Physics. 1992, 159, (1),67-73