Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122
Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122
Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122
Thermo Scientific Chemicals

Palladium, 2.5% Platinum, 2.5% on carbon paste, Type 122

CAS: 7440-05-3 | Pd | 106.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Catalog number H36548.06
also known as H36548-06
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
123.65
Special offer
Online exclusive
Ends: 30-Jun-2026
145.00
Save 21.35 (15%)
Each
Chemical Identifiers
CAS7440-05-3
IUPAC Namepalladium
Molecular FormulaPd
InChI KeyKDLHZDBZIXYQEI-UHFFFAOYSA-N
SMILES[Pd]
View more
SpecificationsSpecification SheetSpecification Sheet
Loss on Drying≤70.0%
Elemental AnalysisPd: 2.25-2.75%
Elemental AnalysisPt: 2.25-2.75%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only