4-Chloro-3-iodo-1H-indazole, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

4-Chloro-3-iodo-1H-indazole, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36741.06
also known as H36741-06
Price (USD)
706.65
Special offer
Online exclusive
Ends: 30-Jun-2026
831.00
Save 124.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
706.65
Special offer
Online exclusive
Ends: 30-Jun-2026
831.00
Save 124.35 (15%)
Each
Chemical Identifiers
CAS518990-33-5
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Cream
FormPowder
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.