Ethyl 2-acetylisonicotinate, 96%
Ethyl 2-acetylisonicotinate, 96%
Ethyl 2-acetylisonicotinate, 96%
Thermo Scientific Chemicals

Ethyl 2-acetylisonicotinate, 96%

CAS: 25028-32-4 | C10H11NO3 | 193.20 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H36911.06
also known as H36911-06
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
286.65
Online Exclusive
318.00
Save 31.35
Each
Chemical Identifiers
CAS25028-32-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale brown
FormCrystals or powder or crystalline powder
Assay (GC)≥95.0%
Melting Point (clear melt)47.0-57.0?C
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only