Azilsartan
Thermo Scientific™

Azilsartan

Have Questions?
Catalog number H37018.03
also known as H37018-03
Price (USD)
465.65
Online Exclusive
517.00
Save 51.35 (10%)
Each
Request bulk or custom format
Price (USD)
465.65
Online Exclusive
517.00
Save 51.35 (10%)
Each
Chemical Identifiers
CAS147403-03-0
IUPAC Name2-ethoxy-1-{[2'-(5-oxo-2,5-dihydro-1,2,4-oxadiazol-3-yl)-[1,1'-biphenyl]-4-yl]methyl}-1H-1,3-benzodiazole-7-carboxylic acid
Molecular FormulaC25H20N4O5
InChI KeyKGSXMPPBFPAXLY-UHFFFAOYSA-N
SMILESCCOC1=NC2=CC=CC(C(O)=O)=C2N1CC1=CC=C(C=C1)C1=CC=CC=C1C1=NC(=O)ON1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream or pale brown
FormCrystals or crystalline powder
Assay (HPLC)≥98.5%
Melting Point (clear melt)200-206?C