Piperazine hexahydrate, 97+%
Piperazine hexahydrate, 97+%
Piperazine hexahydrate, 97+%
Thermo Scientific Chemicals

Piperazine hexahydrate, 97+%

CAS: 142-63-2 | C4H22N2O6 | 194.228 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1000 g
Catalog number H37069.30
also known as H37069-30
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.60
Save 6.95
Each
Quantity:
250 g
Request bulk or custom format
Price (USD)
39.65
Special offer
Online exclusive
Ends: 28-Dec-2026
46.60
Save 6.95
Each
Chemical Identifiers
CAS142-63-2
IUPAC Namepiperazine hexahydrate
Molecular FormulaC4H22N2O6
InChI KeyAVRVZRUEXIEGMP-UHFFFAOYSA-N
SMILESO.O.O.O.O.O.C1CNCCN1
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder or flakes
Assay (Aqueous acid-base Titration)≥97.0 to ≤103.0% (as hexahydrate)
Appearance (Color)Colourless to white
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only