1,3,5-Tris(4-aminophenyl)benzene, 97%
1,3,5-Tris(4-aminophenyl)benzene, 97%
Thermo Scientific Chemicals

1,3,5-Tris(4-aminophenyl)benzene, 97%

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37173.06
also known as H37173-06
Price (USD)
369.65
Online Exclusive
410.00
Save 40.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
369.65
Online Exclusive
410.00
Save 40.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS118727-34-7
IUPAC Name3',5'-bis(4-aminophenyl)-[1,1'-biphenyl]-4-amine
Molecular FormulaC24H21N3
InChI KeyQHQSCKLPDVSEBJ-UHFFFAOYSA-N
SMILESNC1=CC=C(C=C1)C1=CC(=CC(=C1)C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)≥96.0%
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow
1. Key ingredient in a novel ferrocene templated Pd molecular cage catalyst (aka Molecular reactor) used in the synthesis of biphenyls under mild conditions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.