Oxymatrine, 98%
Oxymatrine, 98%
Oxymatrine, 98%
Thermo Scientific™

Oxymatrine, 98%

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H37340.03
also known as H37340-03
Price (USD)
390.65
Online Exclusive
434.00
Save 43.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
390.65
Online Exclusive
434.00
Save 43.35 (10%)
Each
Chemical Identifiers
CAS16837-52-8
IUPAC Name(1R,2R,9S,17S)-6-oxo-7,13-diazatetracyclo[7.7.1.0²,⁷.0¹³,¹⁷]heptadecan-13-ium-13-olate
Molecular FormulaC15H24N2O2
InChI KeyXVPBINOPNYFXID-LHDUFFHYSA-N
SMILES[O-][N+]12CCC[C@H]3CN4[C@H](CCCC4=O)[C@@H](CCC1)[C@@H]23
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale cream
FormPowder
Assay (HPLC)≥97.5%
Melting Point (clear melt)196.0-202.0?C