(2-Hydroxybenzyl)diphenylphosphine oxide, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2-Hydroxybenzyl)diphenylphosphine oxide, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37457.03
also known as H37457-03
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
133.65
Online exclusive
148.00
Save 14.35
Each
Chemical Identifiers
CAS70127-50-3
SpecificationsSpecification SheetSpecification Sheet
Melting Point (clear melt)166.5-172.5?C
Assay (HPLC)≥97.5%
Appearance (Color)White
FormPowder
1. Reagent used in redox neutral organocatalytic Mitsunobu reactions.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.