(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee
(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee
Thermo Scientific Chemicals

(S)-(-)-5,5',6,6',7,7',8,8'-Octahydro-1,1'-bi-2-naphthol, 97+%, 99+% ee

CAS: 65355-00-2 | C20H22O2 | 294.39 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37716.03
also known as H37716-03
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
95.65
Online Exclusive
106.00
Save 10.35 (10%)
Each
Chemical Identifiers
CAS65355-00-2
IUPAC Name5H,5'H,6H,6'H,7H,7'H,8H,8'H-[1,1'-binaphthalene]-2,2'-diol
Molecular FormulaC20H22O2
InChI KeyUTXIFKBYNJRJPH-UHFFFAOYSA-N
SMILESOC1=C(C2=C(CCCC2)C=C1)C1=C(O)C=CC2=C1CCCC2
View more
SpecificationsSpecification SheetSpecification Sheet
Water Content (Karl Fischer Titration)≤0.5%
Melting Point160-168°C
Optical Rotation-48° ± 1.5° (c=1 in chloroform)
Appearance (Color)White to cream
FormPowder
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only