Celite∣r 535
Thermo Scientific Chemicals

Celite∣r 535

CAS: 68855-54-9 | SiO{2}
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
H37758.0B
also known as H37758-0B
1000 g
H37758.0I
also known as H37758-0I
5000 g
Catalog number H37758.0B
also known as H37758-0B
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.90
Save 12.25 (15%)
Each
Quantity:
1000 g
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
83.90
Save 12.25 (15%)
Each
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
CAS68855-54-9
Melting Point>450°C
Molecular FormulaSiO{2}
SynonymDiatomaceous Earth;Filter aid, flux calcined
Quantity1000 g
Unit SizeEach