2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%
2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%
2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%
Thermo Scientific Chemicals

2,2':6',2″-Terpyridine-4'-carboxylic acid, 95%

CAS: 148332-36-9 | C16H11N3O2 | 277.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H37760.MD
also known as H37760-MD
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.50
Save 12.85 (15%)
Each
Quantity:
250 mg
Request bulk or custom format
Price (USD)
75.65
Special offer
Online exclusive
Ends: 30-Jun-2026
88.50
Save 12.85 (15%)
Each
Chemical Identifiers
CAS148332-36-9
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale cream to pale yellow
Assay (Aqueous acid-base Titration)≥94.0 to ≤106.0%
FormPowder
Assay (Silylated GC)≥94.0%

2,2':6',2″-Terpyridine-4'-carboxylic acid is used as dyestuff intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,2′:6′,2′′-Terpyridine-4′-carboxylic acid is used as dyestuff intermediate.

Solubility
Slightly soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
RUO – Research Use Only

General References:

  1. Charles Mikel; Pierre G.Potvin. Synthesis and enhanced photosensitization ability of a 4-carboxy-2,2':6',2″-terpyridine complex of ruthenium(II). Polyhedron. 2002, 21, (1),49-54
  2. Marcel Heller; Ulrich S.Schubert. Syntheses of Functionalized 2,2':6',2″-Terpyridines. European Journal of Organic Chemistry. 2003, 2003, (6),947-961