4'-(4-Bromophenyl)-2,2':6',2″-terpyridine, 97%
4'-(4-Bromophenyl)-2,2':6',2″-terpyridine, 97%
4'-(4-Bromophenyl)-2,2':6',2″-terpyridine, 97%
Thermo Scientific Chemicals

4'-(4-Bromophenyl)-2,2':6',2″-terpyridine, 97%

CAS: 89972-76-9 | C21H14BrN3 | 388.268 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37778.03
also known as H37778-03
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
108.65
Online Exclusive
120.00
Save 11.35 (9%)
Each
Chemical Identifiers
CAS89972-76-9
IUPAC Name4-(4-bromophenyl)-6-(pyridin-2-yl)-2,2'-bipyridine
Molecular FormulaC21H14BrN3
InChI KeyBOPPHKMZOYPASP-UHFFFAOYSA-N
SMILESBrC1=CC=C(C=C1)C1=CC(=NC(=C1)C1=CC=CC=N1)C1=CC=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to pale yellow
Assay from Supplier's CofA≥96.0%
FormCrystals or powder or crystalline powder
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only