1,3,5-Tris(4-methylphenyl)benzene, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,3,5-Tris(4-methylphenyl)benzene, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H37958.03
also known as H37958-03
Price (USD)
103.65
Special offer
Online exclusive
Ends: 28-Dec-2026
122.00
Save 18.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
103.65
Special offer
Online exclusive
Ends: 28-Dec-2026
122.00
Save 18.35
Each
Chemical Identifiers
CAS50446-43-0
SpecificationsSpecification SheetSpecification Sheet
FormCrystals or powder or crystalline powder
Appearance (Color)White to yellow
Assay (GC)≥96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.