3-Amino-4-iodopyridine, 98%
3-Amino-4-iodopyridine, 98%
3-Amino-4-iodopyridine, 98%
Thermo Scientific Chemicals

3-Amino-4-iodopyridine, 98%

CAS: 105752-11-2 | C5H5IN2 | 220.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50023.03
also known as H50023-03
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
153.00
Save 22.35
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
130.65
Special offer
Online exclusive
Ends: 28-Dec-2026
153.00
Save 22.35
Each
Chemical Identifiers
CAS105752-11-2
SpecificationsSpecification SheetSpecification Sheet
Assay (HPLC)97.5% min
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only