7-Azaindole-5-boronic acid pinacol ester, 97%
7-Azaindole-5-boronic acid pinacol ester, 97%
7-Azaindole-5-boronic acid pinacol ester, 97%
7-Azaindole-5-boronic acid pinacol ester, 97%
Thermo Scientific Chemicals

7-Azaindole-5-boronic acid pinacol ester, 97%

CAS: 754214-56-7 | C13H17BN2O2 | 244.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50044.03
also known as H50044-03
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
124.65
Online Exclusive
138.00
Save 13.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS754214-56-7
IUPAC Name5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-1H-pyrrolo[2,3-b]pyridine
Molecular FormulaC13H17BN2O2
InChI KeyUOXAMYZTYZLSCC-UHFFFAOYSA-N
SMILESCC1(C)OB(OC1(C)C)C1=CN=C2NC=CC2=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Assay from Suppliers CofA≥96.0% (HPLC)
Appearance (Color)White to pale yellow
7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
7-Azaindole-5-boronic acid pinacol ester is used as pharmaceutical intermediate.

Solubility
Soluble in methanol, ethanol and THF.

Notes
Air sensitive. Store away from oxidizing agents.
RUO – Research Use Only

General References:

  1. Mark A.Seefeld; Meagan B.Rouse; Kenneth C.McNulty; Lihui Sun; Jizhou Wang; Dennis S.Yamashita; Juan I.Luengo; ShuYun Zhang; Elisabeth A.Minthorn; Nestor O.Concha; Dirk A.Heerding. Discovery of 5-pyrrolopyridinyl-2-thiophenecarboxamides as potent AKT kinase inhibitors. Bioorganic & Medicinal Chemistry Letters. 2009, 19, (8),2244-2248