Specifications
CAS532391-31-4
Chemical Name or Material2-Methoxypyridine-3-boronic acid pinacol ester
Formula Weight235.09
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more