Specifications
CAS362661-83-4
Chemical Name or MaterialPyrazolo[1,5-a]pyridine-7-carboxaldehyde
Formula Weight146.15
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more