2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

2-(Boc-amino)pyridine-5-carboxaldehyde, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50075.03
also known as H50075-03
1 g
Catalog number H50075.03
also known as H50075-03
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
141.65
Special offer
Online exclusive
Ends: 30-Jun-2026
167.00
Save 25.35 (15%)
Each
Chemical Identifiers
CAS199296-40-7
IUPAC Nametert-butyl N-(5-formylpyridin-2-yl)carbamate
Molecular FormulaC11H14N2O3
InChI KeyWZROBBWIJBBWQP-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NC1=NC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
FormSolid
Assay (HPLC)>96.0%
2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(Boc-amino)pyridine-5-carboxaldehyde is used as active pharmaceutical ingredient or intermediates, fine chemicals.

Solubility
Soluble in water.

Notes
Air & Light Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only