2-Ethoxypyridine-3-carboxaldehyde, 97%
2-Ethoxypyridine-3-carboxaldehyde, 97%
2-Ethoxypyridine-3-carboxaldehyde, 97%
Thermo Scientific Chemicals

2-Ethoxypyridine-3-carboxaldehyde, 97%

CAS: 885278-07-9 | C8H9NO2 | 151.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50088.06
also known as H50088-06
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
400.00
Save 59.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
340.65
Special offer
Online exclusive
Ends: 30-Jun-2026
400.00
Save 59.35 (15%)
Each
Chemical Identifiers
CAS885278-07-9
IUPAC Name2-ethoxypyridine-3-carbaldehyde
Molecular FormulaC8H9NO2
InChI KeyWHEFRNNNELKVRQ-UHFFFAOYSA-N
SMILESCCOC1=C(C=O)C=CC=N1
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Appearance (Color)Colorless
Assay (HPLC)>96.0%
Used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used as pharmaceutical intermediate.

Notes
Air Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only