5-Bromo-2-methoxy-3-methylpyridine, 97%
5-Bromo-2-methoxy-3-methylpyridine, 97%
5-Bromo-2-methoxy-3-methylpyridine, 97%
5-Bromo-2-methoxy-3-methylpyridine, 97%
Thermo Scientific Chemicals

5-Bromo-2-methoxy-3-methylpyridine, 97%

CAS: 760207-87-2 | C7H8BrNO | 202.051 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50113.06
also known as H50113-06
Price (USD)
438.65
Special offer
Online exclusive
Ends: 30-Jun-2026
516.00
Save 77.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
438.65
Special offer
Online exclusive
Ends: 30-Jun-2026
516.00
Save 77.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS760207-87-2
IUPAC Name5-bromo-2-methoxy-3-methylpyridine
Molecular FormulaC7H8BrNO
InChI KeyRHYCFMXPPGZHAW-UHFFFAOYSA-N
SMILESCOC1=NC=C(Br)C=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow
FormLiquid
Assay (GC)≥96.0%
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only