1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals
1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals
1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals
1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,6-Naphthyridin-2(1H)-one, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50121.03
also known as H50121-03
1 g
Catalog number H50121.03
also known as H50121-03
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
220.65
Special offer
Online exclusive
Ends: 30-Jun-2026
259.00
Save 38.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS23616-29-7
IUPAC Name1,2-dihydro-1,6-naphthyridin-2-one
Molecular FormulaC8H6N2O
InChI KeyZQKMVHXJWJNEQG-UHFFFAOYSA-N
SMILESO=C1NC2=CC=NC=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Pale yellow to brown
Assay from Suppliers CofA≥96.0%
FormCrystals or powder or crystalline powder
Proton NMRConforms to structure
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only