2-(2-Pyrrolidinyl)pyridine, 96%
2-(2-Pyrrolidinyl)pyridine, 96%
2-(2-Pyrrolidinyl)pyridine, 96%
Thermo Scientific Chemicals

2-(2-Pyrrolidinyl)pyridine, 96%

CAS: 77790-61-5 | C9H12N2 | 148.209 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50176.03
also known as H50176-03
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
242.65
Online Exclusive
269.00
Save 26.35 (10%)
Each
Specifications
CAS77790-61-5
Chemical Name or Material2-(2-Pyrrolidinyl)pyridine
DOT InformationTransport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
Formula Weight148.21
Health Hazard 1H301+H311+H331-H315-H319-H335
View more
2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-(2-Pyrrolidinyl)pyridine is used as primary and secondary intermediate.

Solubility
Soluble in water.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only

General References:

  1. Giorgio Chelucci.; Vittorio Caria.; Antonio Saba. Enantioselective palladium catalyzed allylic substitution with chiral pyridine ligands. Journal of Molecular Catalysis A: Chemical. 1998, 130, (1-2), 51-55.
  2. David F. Glenn.; William B. EdwardsIII. Synthesis and mass spectrometry of some structurally related nicotinoids. J. Org. Chem. 1978, 43, (14), 2860-2870.