3-(4-Chlorophenyl)propionic acid, 94%
3-(4-Chlorophenyl)propionic acid, 94%
3-(4-Chlorophenyl)propionic acid, 94%
Thermo Scientific Chemicals

3-(4-Chlorophenyl)propionic acid, 94%

CAS: 2019-34-3 | C9H9ClO2 | 184.619 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50204.03
also known as H50204-03
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
40.65
Special offer
Online exclusive
Ends: 30-Jun-2026
47.60
Save 6.95 (15%)
Each
Chemical Identifiers
CAS2019-34-3
IUPAC Name3-(4-chlorophenyl)propanoic acid
Molecular FormulaC9H9ClO2
InChI KeyBBSLOKZINKEUCR-UHFFFAOYSA-N
SMILESOC(=O)CCC1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to brown
FormPowder or crystals or crystalline powder
Assay from Supplier's CofA≥92.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only