3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

3-Bromo-2-fluorobenzoic acid, 97%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50235.06
also known as H50235-06
5 g
Catalog number H50235.06
also known as H50235-06
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
221.65
Online Exclusive
246.00
Save 24.35 (10%)
Each
Chemical Identifiers
CAS161957-56-8
IUPAC Name3-bromo-2-fluorobenzoic acid
Molecular FormulaC7H4BrFO2
InChI KeyUVKURTLVTLRSSM-UHFFFAOYSA-N
SMILESOC(=O)C1=C(F)C(Br)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
FormSolid
Appearance (Color)Off-white
Assay (HPLC)>96.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only