4-(Methylsulfonyl)benzenesulfonyl chloride, 95%
4-(Methylsulfonyl)benzenesulfonyl chloride, 95%
4-(Methylsulfonyl)benzenesulfonyl chloride, 95%
4-(Methylsulfonyl)benzenesulfonyl chloride, 95%
Thermo Scientific Chemicals

4-(Methylsulfonyl)benzenesulfonyl chloride, 95%

CAS: 82964-91-8 | C7H7ClO4S2 | 254.70 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50264.06
also known as H50264-06
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
206.65
Online Exclusive
229.00
Save 22.35 (10%)
Each
Add to cart
Chemical Identifiers
CAS82964-91-8
IUPAC Name4-methanesulfonylbenzene-1-sulfonyl chloride
Molecular FormulaC7H7ClO4S2
InChI KeyTYJOQICPGZGYDT-UHFFFAOYSA-N
SMILESCS(=O)(=O)C1=CC=C(C=C1)S(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Assay from Suppliers CofA≥94.0%
FormSolid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only