N,N-Diethyl-1,4-butanediamine, 96%
N,N-Diethyl-1,4-butanediamine, 96%
N,N-Diethyl-1,4-butanediamine, 96%
N,N-Diethyl-1,4-butanediamine, 96%
Thermo Scientific Chemicals

N,N-Diethyl-1,4-butanediamine, 96%

CAS: 27431-62-5 | C8H20N2 | 144.262 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50278.06
also known as H50278-06
Price (USD)
443.65
Special offer
Online exclusive
Ends: 30-Jun-2026
522.00
Save 78.35 (15%)
Each
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)
443.65
Special offer
Online exclusive
Ends: 30-Jun-2026
522.00
Save 78.35 (15%)
Each
Add to cart
Chemical Identifiers
CAS27431-62-5
IUPAC Name(4-aminobutyl)diethylamine
Molecular FormulaC8H20N2
InChI KeyJILXUIANNUALRZ-UHFFFAOYSA-N
SMILESCCN(CC)CCCCN
View more
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay by NMR>95.0%
Appearance (Color)Colorless
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N,N-Diethyl-1,4-butanediamine is used as pharmaceutical intermediate.

Solubility
Slightly soluble in water.

Notes
Air and moisture sensitive. Incompatible with oxidizing agents.
RUO – Research Use Only
Raymond J.Bergeron; Allen H.Neims; James S.McManis; Thomas R.Hawthorne; John R.T.Vinson; Rita Bortell; Michael J.Ingeno. Synthetic polyamine analogs as antineoplastics. J. Med. Chem. 1988, 31, (6),1183-1190