(2,6-Dichlorophenylthio)acetic acid, 99%
(2,6-Dichlorophenylthio)acetic acid, 99%
(2,6-Dichlorophenylthio)acetic acid, 99%
Thermo Scientific Chemicals

(2,6-Dichlorophenylthio)acetic acid, 99%

CAS: 21248-45-3 | C8H5Cl2O2S | 236.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50282.03
also known as H50282-03
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
121.65
Online Exclusive
135.00
Save 13.35 (10%)
Each
Chemical Identifiers
CAS21248-45-3
IUPAC Name2-[(2,6-dichlorophenyl)sulfanyl]acetate
Molecular FormulaC8H5Cl2O2S
InChI KeyURNRRSJGOOTQBO-UHFFFAOYSA-M
SMILES[O-]C(=O)CSC1=C(Cl)C=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
FormCrystalline solid
Assay (HPLC)>98.5%
Appearance (Color)White
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only