N-Methyl(diphenylmethyl)amine, 98%
N-Methyl(diphenylmethyl)amine, 98%
N-Methyl(diphenylmethyl)amine, 98%
Thermo Scientific Chemicals

N-Methyl(diphenylmethyl)amine, 98%

CAS: 14683-47-7 | C14H15N | 197.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50327.03
also known as H50327-03
Price (USD)
51.30
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
51.30
Each
Specifications
Beilstein2096409
CAS14683-47-7
Chemical Name or MaterialN-Methyl(diphenylmethyl)amine
Formula Weight197.28
Health Hazard 1H315-H319-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only