(R)-(-)-2-Benzylamino-1-butanol, 99%, Thermo Scientific Chemicals
(R)-(-)-2-Benzylamino-1-butanol, 99%, Thermo Scientific Chemicals
(R)-(-)-2-Benzylamino-1-butanol, 99%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(R)-(-)-2-Benzylamino-1-butanol, 99%, Thermo Scientific Chemicals

Have Questions?
Catalog NumberQuantity
H50328.03
also known as H50328-03
1 g
Catalog number H50328.03
also known as H50328-03
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
111.65
Special offer
Online exclusive
Ends: 30-Jun-2026
131.00
Save 19.35 (15%)
Each
Specifications
CAS6257-49-4
Chemical Name or Material(R)-(-)-2-Benzylamino-1-butanol
Formula Weight179.26
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only