2,4,6-Trimethoxybenzyl alcohol, 97%
2,4,6-Trimethoxybenzyl alcohol, 97%
2,4,6-Trimethoxybenzyl alcohol, 97%
2,4,6-Trimethoxybenzyl alcohol, 97%
Thermo Scientific Chemicals

2,4,6-Trimethoxybenzyl alcohol, 97%

CAS: 61040-78-6 | C10H14O4 | 198.218 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 mg
1 g
Catalog number H50348.03
also known as H50348-03
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)
177.65
Online Exclusive
197.00
Save 19.35 (10%)
Each
Add to cart
Specifications
CAS61040-78-6
Chemical Name or Material2,4,6-Trimethoxybenzyl alcohol
Formula Weight198.22
IUPAC Name(2,4,6-trimethoxyphenyl)methanol
InChI KeyCFXXBVNHYJQNKS-UHFFFAOYSA-N
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only