Specifications
CAS17243-13-9
Chemical Name or Material5-(Chlorosulfonyl)salicylic acid
DOT InformationTransport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
Formula Weight236.63
Health Hazard 1H314-H335
View more