Specifications
CAS6461-76-3
Chemical Name or Material1,2-Benzenedisulfonyl chloride
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
Formula Weight275.13
Health Hazard 1Danger
View more