(Methylsulfonyl)methanesulfonyl chloride, 98%
(Methylsulfonyl)methanesulfonyl chloride, 98%
(Methylsulfonyl)methanesulfonyl chloride, 98%
Thermo Scientific Chemicals

(Methylsulfonyl)methanesulfonyl chloride, 98%

CAS: 22317-89-1 | C2H5ClO4S2 | 192.628 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50385.06
also known as H50385-06
Price (USD)
423.65
Special offer
Online exclusive
Ends: 30-Jun-2026
498.00
Save 74.35 (15%)
Each
Quantity:
5 g
Request bulk or custom format
Price (USD)
423.65
Special offer
Online exclusive
Ends: 30-Jun-2026
498.00
Save 74.35 (15%)
Each
Chemical Identifiers
CAS22317-89-1
IUPAC Namemethanesulfonylmethanesulfonyl chloride
Molecular FormulaC2H5ClO4S2
InChI KeyDQDYGBSUPOHHMW-UHFFFAOYSA-N
SMILESCS(=O)(=O)CS(Cl)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Assay (unspecified)>97.5%
FormPowder
(Methylsulfonyl)methanesulfonyl chloride is used as a chemicals, pharmaceutical intermediates, reagents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
(Methylsulfonyl)methanesulfonyl chloride is used as a chemicals, pharmaceutical intermediates, reagents.

Solubility
Soluble in water (50g/L).

Notes
Moisture Sensitive & Hygroscopic. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
RUO – Research Use Only
Y. Fulmer Shealy.; Charles A. Krauth.; W. Russell LasterJr.. 2-Chloroethyl (methylsulfonyl)methanesulfonate and related (methylsulfonyl)methanesulfonates. Antineoplastic activity in vivo. J. Med. Chem. 1984, 27, (5), 664-670.