1-Naphthylacetyl chloride, 95%
1-Naphthylacetyl chloride, 95%
1-Naphthylacetyl chloride, 95%
Thermo Scientific Chemicals

1-Naphthylacetyl chloride, 95%

CAS: 5121-00-6 | C12H9ClO | 204.65 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50386.03
also known as H50386-03
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
87.65
Special offer
Online exclusive
Ends: 30-Jun-2026
103.00
Save 15.35 (15%)
Each
Chemical Identifiers
CAS5121-00-6
IUPAC Name2-(naphthalen-1-yl)acetyl chloride
Molecular FormulaC12H9ClO
InChI KeyDSVAZLXLRDXHKO-UHFFFAOYSA-N
SMILESClC(=O)CC1=CC=CC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Yellow
FormLiquid
Assay (unspecified)≥94.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only