8-Hydroxyquinoline-2-carbonitrile, 98%
8-Hydroxyquinoline-2-carbonitrile, 98%
8-Hydroxyquinoline-2-carbonitrile, 98%
Thermo Scientific Chemicals

8-Hydroxyquinoline-2-carbonitrile, 98%

CAS: 6759-78-0 | C10H6N2O | 170.171 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number H50433.03
also known as H50433-03
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.30
Save 12.65 (15%)
Each
Quantity:
1 g
Request bulk or custom format
Price (USD)
71.65
Special offer
Online exclusive
Ends: 30-Jun-2026
84.30
Save 12.65 (15%)
Each
Chemical Identifiers
CAS6759-78-0
IUPAC Name8-hydroxyquinoline-2-carbonitrile
Molecular FormulaC10H6N2O
InChI KeyKUQKKIBQVSFDHX-UHFFFAOYSA-N
SMILESOC1=C2N=C(C=CC2=CC=C1)C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)>97.5%
8-Hydroxyquinoline-2-carbonitrile is a building block for preparing chelating agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
8-Hydroxyquinoline-2-carbonitrile is a building block for preparing chelating agents.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
RUO – Research Use Only
Christina Moberg; Mamoun Muhammed; Göran Svensson and Michael Weber. Novel quinaldic acids for selective chelation of cadmium(II). X-Ray crystal structure of [Cd(C18H12NO4)2](Me2SO)·2H2O. J. Chem. Soc., Chem. Commun.1988, 810-812.